Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TER-119 Monoclonal Antibody (TER-119), Biotin, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

$104.00 - $315.00

Specifications

Antigen TER-119
Clone TER-119
Concentration 0.5 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-120-40
Encompass_Preferred
View Documents
Invitrogen™
13592181
50 μg
Each for $104.00
Only null left
Edge
Add to Cart
 
50-112-9513
Encompass_Preferred
View Documents
Invitrogen™
13592182
100 μg
Each for $199.00
Only null left
Edge
Add to cart
 
50-120-42
Encompass_Preferred
View Documents
Invitrogen™
13592185
500 μg
Each for $315.00
Only null left
Add to cart
 
Description

Description

Description: The TER-119 monoclonal antibody reacts with mouse erythroid cells from early proerythroblast to mature erythrocyte stages. The TER-119 antigen is present in yolk sac, fetal and newborn liver, but is not expressed by cells carrying BFU-E and CFU-E activities. Several erythroleukemia cell lines tested so far are negative for expression of TER-119 antigen even after dimethylsulfoxide stimulation. Biochemical and molecular analysis of the TER-119 antigen indicate that this molecule is associated with the surface glycophorin A, but is not a typical glycophorin. Applications Reported: The TER-119 antibody has been reported for use in flow cytometric analysis. Applications Tested: The TER-119 antibody has been tested by flow cytometric analysis of mouse splenocytes and bone marrow cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.

TER-119 is a lineage marker for erythroid cells from early proerythroblast to mature erythrocyte stages in adult blood, spleen, and bone marrow. It is also present in yolk sac, and fetal and newborn liver. The TER-119 antigen is not expressed by cells of earlier erythroid development at BFU-e (blast-forming unit erythroid) stage or CFU-e (colony-forming unit erythroid) stage.
Specifications

Specifications

TER-119
0.5 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
Ly76; Ly-76; TER119
Ly76
Primary
4°C, store in dark, DO NOT FREEZE!
Ly76
TER-119
Flow Cytometry
Biotin
Rat
Mouse
0
104231
IgG2b κ
Affinity chromatography
Antibody
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

Product Certifications

For Research Use Only.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.