Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Terbinafine hydrochloride
SDP

Catalog No. 648450
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-845-0 50 mg
64-842-50 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 64-845-0 Supplier Tocris Bioscience™ Supplier No. 6484/50

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective TASK3 activator

Terbinafine hydrochloride is a selective TASK3 activator (pEC50 = 6.2). Exhibits >10-fold selectivity for TASK3 over TREK2, TRESK, THIK1 and TASK2. Also inhibits TWIK1 (pIC50 = 5.69). Also antifungal; inhibits fungal sterol biosynthesis by inhibition of squalene epoxidase.

Specifications

CAS 78628-80-5
Synonym N-[(2E)-6,6-Dimethyl-2-hepten-4-yn-1-yl]-N-methyl-1-naphthalenemethanamine hydrochloride
Quantity 50 mg
Molecular Formula C21H25N.HCl
Formula Weight 327.89
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.