Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terbinafine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog No. AC461260010
Click to view available options
Quantity:
1 g
5 g

Catalog No. AC461260010

Supplier: Thermo Scientific Chemicals 461260010

Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78628-80-5
Molecular Formula C21H25N·HCl
Molecular Weight (g/mol) 327.89
InChI Key BWMISRWJRUSYEX-SZKNIZGXSA-N
Synonym terbinafine hydrochloride, terbinafine hcl, lamisil, terbinafine hydrochioride, lamisil krem, lamisil at, zabel, terbinafinum latin, terbinafina spanish, zimig
PubChem CID 5282481
ChEBI CHEBI:77614
IUPAC Name (E)-N,6,6-trimethyl-N-(naphthalen-1-ylmethyl)hept-2-en-4-yn-1-amine;hydrochloride
SMILES CC(C)(C)C#CC=CCN(C)CC1=CC=CC2=CC=CC=C21.Cl

Specifications

Melting Point 214.0°C
Loss on Drying 0.5% max. (1 g, 105)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information (5% in methanol) Clear solution
Formula Weight 327.89
Percent Purity 99%
Chemical Name or Material Terbinafine hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement Avoid release to the environment. Collect spillage. Dispose of contents/container to an approved waste disposal plant.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.