Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terbium(III) chloride hexahydrate, 99.9%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
13798-24-8
Cl3Tb·6H2O
373.38
MFCD00149870
13798-24-8
Cl3Tb·6H2O
373.38
MFCD00149870
for ultra trace analysis
Assay Percent Range 99.90%
Linear Formula TbCl3·6H2O
Merck Index 14, 9157
Solubility Information Solubility in water: soluble. Other solubilities: readily soluble in alcohol
Formula Weight 373.38
Physical Form Crystalline Powder, Crystals, and/or Chunks
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Solubility Solubility in water: soluble. Other solubilities: readily soluble in alcohol
Terbium (Tb) 42.13 to 42.99%
Total Impurity 0.1% max.
Color White
Quantity 5g
Chemical Name or Material Terbium(III) chloride hexahydrate
Show More Show Less