Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terbium(III,IV) oxide, 99.9% (REO), Alfa Aesar™, Reacton™

Manufacturer:  Alfa Aesar 1120709

Quantity 10g

 View more versions of this product

Catalog No. AA1120709


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

12037-01-3
99.9% (REO)
747.69
Terbium(III, IV) oxide
Powder
Tb4O7
MFCD00083151

Specifications

12037-01-3
99.9% (REO)
747.69
10g
Powder
Tb4O7
MFCD00083151
Terbium(III, IV) oxide

Safety and Handling

SDS & Safety Documents

EINECSNumber : 234-856-3

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit