Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terbium(III) nitrate hexahydrate, 99.99+%, (trace metal basis)

Catalog No. p-221660
Click to view available options
Quantity:
2 g
10 g

CAS: 13451-19-9 | H12N3O15Tb | 453.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13451-19-9
Molecular Formula H12N3O15Tb
Molecular Weight (g/mol) 453.03
MDL Number MFCD00799162,MFCD00149868
InChI Key UTCARTSNNKGRTD-UHFFFAOYSA-N
Synonym terbium trinitrate hexahydrate, terbium iii nitrate hexahydrate, terbium nitrate, hexahydrate, terbium iii nitrate, hexahydrate 1:3:6, terbium nitrate hexahydrate, nitric acid, terbium 3+ salt, hexahydrate, 3no3.tb.6h2o, terbium iii nitratehexahydrate, terbium 3+ trinitrate hexahydrate, terbium iii nitrate hydrate, reacton
PubChem CID 202879
IUPAC Name terbium(3+);trinitrate;hexahydrate
SMILES O.O.O.O.O.O.[Tb+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color White
Physical Form Granular Powder
Quantity 2 g
Assay Percent Range 99.99% min. (TREO base)
Linear Formula Tb(NO3)3·6H2O
Beilstein 11,9087
Merck Index 14,9157
Solubility Information Solubility in water: soluble
Formula Weight 453.02
Percent Purity ≥99.99%
Grade Trace Metal Basis
Chemical Name or Material Terbium(III) nitrate hexahydrate, 99.9%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Take any precaution to avoid mixing with combustibles.
RTECSNumber WY9625000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.