Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terbium(III) nitrate hydrate, 99.9% (REO)

Catalog No. p-4408186
Click to view available options
Quantity:
5 g
25 g

Terbium(III) nitrate hydrate, H2N3O10Tb, CAS Number-94219-56-4, terbium 3+ hydrate trinitrate, 3no3.tb.h2o, 25g, 99.9% (REO), Hazard Class: 5.1; Packaging Group: III, 233-138-7, 344.94 (anhydrous), H272-H315-H319-H335 | CAS: 94219-56-4 | N3O9Tb | 344.94 g/mol

Terbium(III) nitrate hexahydrate forms a microporous material, which is used for sensor applications due to its luminescent properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94219-56-4
Molecular Formula N3O9Tb
Molecular Weight (g/mol) 344.94
MDL Number MFCD00011257
InChI Key YJVUGDIORBKPLC-UHFFFAOYSA-N
Synonym 3no3.tb.h2o, terbium 3+ hydrate trinitrate
PubChem CID 91886285
IUPAC Name terbium(3+);trinitrate;hydrate
SMILES [Tb+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Physical Form Wet Crystalline Aggregates
Quantity 5 g
Assay Percent Range 99.9% (REO)
Solubility Information Soluble in water.
Formula Weight 344.94 (Anhydrous)
Chemical Name or Material Terbium(III) nitrate hydrate
Hazard Category H272-H315-H319-H335
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: III
EINECSNumber 233-138-7
RTECSNumber QV1800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.