Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terbium(III) oxalate decahydrate, REacton™, 99.9% (REO), Thermo Scientific™

Catalog No. AA1459704
Click to view available options
Quantity:
2 g

Catalog No. AA1459704

Supplier: Thermo Scientific Chemicals 1459704

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51373-66-1
Molecular Formula C6O12Tb2
Molecular Weight (g/mol) 581.905
MDL Number MFCD00748140
InChI Key JZNKXIAEFQDQDF-UHFFFAOYSA-H
Synonym terbium iii oxalate, diterbium trioxalate, edioic acid, terbium 3 salt 3:2, oxalate; terbium 3+, ethanedioic acid, terbium 3+ salt 3:2, acmc-1cs38, terbium, mu-ethanedioato 2--kappao1,kappao2':kappao1',kappao2 bis ethanedioato 2--kappao1,kappao2 di
PubChem CID 164770
IUPAC Name oxalate;terbium(3+)
SMILES C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].[Tb+3].[Tb+3]

Specifications

Physical Form Crystalline
Quantity 2 g
Linear Formula Tb2(C2O4)3·10H2O
UN Number UN3288
Formula Weight 762.06 (581.91 Anhydrous)
Percent Purity 99.9%
Grade Reacton
Assay (REO)
Chemical Name or Material Terbium(III) oxalate decahydrate
Hazard Category H302+H312
Hazard Statement GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 213-632-9
TSCA Yes
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.