Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terbium(III) perchlorate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific Chemicals

Catalog No. p-7051154
Click to view available options
Quantity:
10 g

In observation of slow magnetic relaxation in triple-stranded lanthanide helicates Terbium(III) perchlorate is substituted for dysprosium(III) perchlorate hexahydrate. By reacting 3-formylchromone and hydrated terbium (III) perchlorate in isopropanol, 3-hydroxymethylchromone [icm], bis(3-chromonyl)methanol [dcm] and the novel complex Tb(imc)3(ClO4)3.5H2O are obtained.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14014-09-6
MDL Number MFCD00150362

Specifications

Color Colorless
Physical Form Liquid
Quantity 10 g
Linear Formula Tb(ClO4)3·6H2O
UN Number UN3211
Solubility Information Insoluble in water.
Formula Weight 565.37 (457.28 Anhydrous)
Grade Reagent
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Odor Odorless
Density 1.5 g/mL
Chemical Name or Material Terbium(III) perchlorate
Show More Show Less
Hazard Category H271-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, AQUEOUS SOLUTION, N.O.S.
EINECSNumber 237-826-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.