Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terbutaline Sulfate, USP, 98-101%, Spectrum™ Chemical
SDP

Catalog No. 18607245 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle

T1062, 23031-32-5, (C12H19NO3)2·H2SO4

Spectrum™ Chemical Terbutaline Sulfate, USP is used in the management of asthma symptoms as a reliever inhaler. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 23031-32-5
Molecular Formula C24H40N2O10S
Molecular Weight (g/mol) 548.65
InChI Key KFVSLSTULZVNPG-UHFFFAOYNA-N
IUPAC Name bis(5-[2-(tert-butylamino)-1-hydroxyethyl]benzene-1,3-diol); sulfuric acid
SMILES OS(O)(=O)=O.CC(C)(C)NCC(O)C1=CC(O)=CC(O)=C1.CC(C)(C)NCC(O)C1=CC(O)=CC(O)=C1

Specifications

CAS Max % 1
Ignition Residue 0.002
Loss on Drying 0.005
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 98 to 101%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.