Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBIAD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA14378
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA14378 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA14378 Supplier Invitrogen™ Supplier No. PA14378
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-4378 detects the TERE1 protein in human samples. PA1-4378 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~37 kDa protein representing TERE1 in urothelial carcinoma. The PA1- 4378 immunogen is a synthetic peptide corresponding to residues P(32) E Q D R L P Q R S W R Q K(45) of human TERE1. There is 85% sequence identity to mouse and rat.

A novel gene designated TERE1, has a significant effect on the growth regulation in bladder cancer. The TERE1 gene maps to a chromosome locus that has been identified by loss of heterozygosity studies as a site of a putative tumor suppressor gene or genes for multiple tumor types including prostate carcinoma. Data suggest that TERE1 may be significant in prostate cancer growth regulation and the down regulation or absence of TERE1 may be an important component of the phenotype of advanced disease. Data also suggest a potential role for this gene product in the progression of bladder cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBIAD1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene UBIAD1
Gene Accession No. Q9Y5Z9
Gene Alias 1200002M06Rik; AI426463; AW320947; RGD1309588; RP4-796F18.1; SCCD; Tere1; transitional epithelia response protein; transitional epithelial response protein 1; UbiA prenyltransferase domain containing 1; ubiA prenyltransferase domain-containing protein 1; UBIAD1
Gene Symbols UBIAD1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues P(32) E Q D R L P Q R S W R Q K(45) of human TERE1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29914
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.