Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBIAD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583743
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583743 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583743 Supplier Invitrogen™ Supplier No. PA583743
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAASQVLGEK INILSGETVK AGDRDPLGND CPEQDRLPQR SWRQKCASYV LALRPWSFSA SLTPVALGSA LAYRSHGVLD PR Highest antigen sequence indentity to the following orthologs: Rat - 80%, Mouse - 82%.

A novel gene designated TERE1, has a significant effect on the growth regulation in bladder cancer. The TERE1 gene maps to a chromosome locus that has been identified by loss of heterozygosity studies as a site of a putative tumor suppressor gene or genes for multiple tumor types including prostate carcinoma. Data suggest that TERE1 may be significant in prostate cancer growth regulation and the down regulation or absence of TERE1 may be an important component of the phenotype of advanced disease. Data also suggest a potential role for this gene product in the progression of bladder cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBIAD1
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UBIAD1
Gene Accession No. Q9Y5Z9
Gene Alias 1200002M06Rik; AI426463; AW320947; RGD1309588; RP4-796F18.1; SCCD; Tere1; transitional epithelia response protein; transitional epithelial response protein 1; UbiA prenyltransferase domain containing 1; ubiA prenyltransferase domain-containing protein 1; UBIAD1
Gene Symbols UBIAD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UBIAD1. Recombinant protein control fragment (Product #RP-104798).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29914
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.