Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terephthaldicarboxaldehyde, 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Plastic bottle
623-27-8
C8H6O2
134.13
MFCD00006949
KUCOHFSKRZZVRO-UHFFFAOYSA-N
1,4-benzenedicarboxaldehyde,p-phthalaldehyde,terephthaldehyde,terephthaldialdehyde,terephthaladehyde,terephthaldicarboxaldehyde,terephthalic aldehyde,1,4-phthalaldehyde,p-formylbenzaldehyde,terephtaldehyde
12173
terephthalaldehyde
C1=CC(=CC=C1C=O)C=O
623-27-8
C8H6O2
134.13
MFCD00006949
KUCOHFSKRZZVRO-UHFFFAOYSA-N
1,4-benzenedicarboxaldehyde,p-phthalaldehyde,terephthaldehyde,terephthaldialdehyde,terephthaladehyde,terephthaldicarboxaldehyde,terephthalic aldehyde,1,4-phthalaldehyde,p-formylbenzaldehyde,terephtaldehyde
12173
terephthalaldehyde
C1=CC(=CC=C1C=O)C=O
Boiling Point 245°C to 248°C
Flash Point 76°C
Melting Point 114°C to 116°C
Color White to Yellow
Quantity 100g
Packaging Plastic bottle
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula C6H4(CHO)2
Beilstein 07, 675
Solubility Information Solubility in water: 3g/L (50°C). Other solubilities: soluble in alkalies,soluble in alcohol and ether
Formula Weight 134.13
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material Terephthaldicarboxaldehyde, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 210-784-8

RTECSNumber : WZ0430000

TSCA : TSCA