Learn More
Terephthalic acid, 98+%
Description
Terephthalic acid is used in the metal organic frame works. Terephthalic acid is used primarily to make polyethylene terephthalate (PET) resins and fibers, polyester fibers, films, polyethylene terephthalate solid-state resins and polyethylene terephthalate engineering resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Terephthalic acid is used in the metal organic frame works. Terephthalic acid is used primarily to make polyethylene terephthalate (PET) resins and fibers, polyester fibers, films, polyethylene terephthalate solid-state resins and polyethylene terephthalate engineering resins.
Solubility
slightly soluble in water (0,017 g/L at 25°C)
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It should be stored and used in molten form.
            Specifications
Specifications
| Melting Point | 402°C (sublimation) | 
| Density | 1.51 | 
| Flash Point | 260°C (500°F) | 
| Odor | Odorless | 
| Quantity | 250 g | 
| Beilstein | 1909333 | 
| Merck Index | 14,9162 | 
| Solubility Information | slightly soluble in water (0,017g/L at 25°C) | 
| Formula Weight | 166.13 | 
| Percent Purity | ≥98% | 
| Show More | 
Safety and Handling
| Hazard Category | H315-H320-H335-H500 | 
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.  | 
                                
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c | 
| EINECSNumber | 202-830-0 | 
| RTECSNumber | WZ0875000 | 
| TSCA | Yes | 
| Recommended Storage | Ambient temperatures | 
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.