Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terephthalic acid, 99+%

Catalog No. p-3758104
Click to view available options
Quantity:
250 g
500 g
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle

CAS: 100-21-0 | C8H6O4 | 166.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-21-0
Molecular Formula C8H6O4
Molecular Weight (g/mol) 166.13
MDL Number MFCD00002558
InChI Key KKEYFWRCBNTPAC-UHFFFAOYSA-N
Synonym p-phthalic acid, 1,4-benzenedicarboxylic acid, benzene-1,4-dicarboxylic acid, p-dicarboxybenzene, p-benzenedicarboxylic acid, p-carboxybenzoic acid, acide terephtalique, para-phthalic acid, tephthol, 1,4-dicarboxybenzene
PubChem CID 7489
ChEBI CHEBI:15702
IUPAC Name terephthalic acid
SMILES C1=CC(=CC=C1C(=O)O)C(=O)O

Specifications

Melting Point >300.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Quantity 250 g
Beilstein 09,841
Merck Index 15,9305
Sublimation Point 0402.0°C
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alkalies, slightly soluble in alcohol, insoluble in chloroform, ether and acetic acid
Formula Weight 166.13
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Terephthalic acid
Show More Show Less
EINECSNumber 202-830-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.