Alfa Aesar™ Terephthalonitrile, 98%

Manufacturer: Alfa Aesar™ B2313714

Quantity 25g

 View more versions of this product

Catalog No. AAB2313714

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 623-26-7
    Molecular Formula C8H4N2
    Formula Weight 128.13
    MDL Number MFCD00001810
    Beilstein 1072210
    Synonym 1,4-Dicyanobenzene; Benzene-1,4-dicarbonitrile
    Chemical Name or Material Terephthalonitrile

    Specifications

    Formula Weight 128.13
    TSCA TSCA
    Molecular Formula C8H4N2
    MDL Number MFCD00001810
    CAS 623-26-7
    Synonym 1,4-Dicyanobenzene; Benzene-1,4-dicarbonitrile
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 224° to 227°C
    Odor Odorless
    Quantity 25g
    RTECS Number CZ1925000
    Beilstein 1072210
    Chemical Name or Material Terephthalonitrile
    Assay 98%

    Description

    Applications
    1,4-dicyanobenzene as a new internal standard in utilizing solid-state infrared intensity methods for the determination of thiocyanate bonding. Used as photosensitizer in [2+2] cycloadditions, nitrile building block.

    Solubility
    Insoluble in water.

    Notes
    Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit