Alfa Aesar™ Terephthaloyl chloride, 99%

Manufacturer: Alfa Aesar™ A1122430

Quantity 250g

 View more versions of this product

Catalog No. AAA1122430

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 100-20-9
    Molecular Formula C8H4Cl2O2
    Formula Weight 203.02
    MDL Number MFCD00000693
    UN Number UN2928
    Beilstein 607796
    Synonym Benzene-1,4-dicarbonyl chloride
    Chemical Name or Material Terephthaloyl chloride

    Specifications

    Formula Weight 203.02
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Health Hazard 1 Danger
    RTECS Number WZ1797000
    Quantity 250g
    Molecular Formula C8H4Cl2O2
    MDL Number MFCD00000693
    Beilstein 607796
    Health Hazard 2 GHS H Statement
    H331-H314-H318
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Boiling Point 265°C
    Melting Point 79° to 83°C
    Flash Point 180°C (356°F)
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S.
    CAS 100-20-9
    UN Number UN2928
    Synonym Benzene-1,4-dicarbonyl chloride
    Chemical Name or Material Terephthaloyl chloride
    Assay 99%

    Description

    Applications
    Terephthaloyl chloride acts as a monomer with p-phenylenediamine used in the preparation of polymer viz. poly paraphenylene terephthalamide. It is an active component in performance polymers and aramid fibers, which gives flame resistance, chemical resistance, temperature stability, light weight, and very high strength. It is also used as an effective water scavenger, utilized to stabilize isocyanates and urethane prepolymers. Further, it is used in the preparation of liquid crystalline thermosets by thermal cyclotrimerization of dicyanate compounds of ring substituted bis(4-hydroxyphenyl) terepthalates. In addition to this, it is involved in the condensation reaction with difunctional alfa, µ-diaminopolystyrene to get chain-extended polystyrene containing amide bonds along the polymer backbone.

    Solubility
    Soluble in ethanol, ether and other common organic solvents.

    Notes
    Incompatible with alcohols, oxidizing agents, water and strong bases.

    GHS H Statement
    H331-H314-H318
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit