Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TERF2IP Antibody (78B356.1), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056526JF646
Description
TERF2IP Monoclonal antibody specifically detects TERF2IP in Human samples. It is validated for Western BlotSpecifications
TERF2IP | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
78B356.1 | |
Western Blot | |
dopamine receptor interacting protein 5, Dopamine receptor-interacting protein 5, DRIP5, hRap1, RAP1 homolog, RAP1TERF2-interacting telomeric protein 1, Repressor/activator protein 1 homolog, telomeric repeat binding factor 2, interacting protein, telomeric repeat-binding factor 2-interacting protein 1, TRF2-interacting telomeric protein 1, TRF2-interacting telomeric RAP1 protein | |
A synthetic peptide corresponding amino acids 212-235 of human Rap1 was used to immunize mice. | |
0.1 mL | |
Cellular Markers, Chromatin Research, DNA Repair, Endosome Markers, Lysosome Markers | |
54386 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction