Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TERF2IP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP18243325UL
Description
TERF2IP Polyclonal specifically detects TERF2IP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TERF2IP | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
dopamine receptor interacting protein 5, Dopamine receptor-interacting protein 5, DRIP5, hRap1, RAP1 homolog, RAP1TERF2-interacting telomeric protein 1, Repressor/activator protein 1 homolog, telomeric repeat binding factor 2, interacting protein, telomeric repeat-binding factor 2-interacting protein 1, TRF2-interacting telomeric protein 1, TRF2-interacting telomeric RAP1 protein | |
Rabbit | |
Affinity Purified | |
RUO | |
54386 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
TERF2IP | |
This antibody was developed against Recombinant Protein corresponding to amino acids:REFEEVVVDESPPDFEIHITMCDDDPPTPEEDSETQPDEEEEEEEEKVSQPEVGAAIKIIRQLMEKFNLDLSTVTQAFLKNSGELEATSAFLASGQRADGYPIWSRQDDIDLQKDDEDTREALVKKFGAQNVAR | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction