Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TERF2IP Antibody (RAP1 5G7/1), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB12014403AF350
Description
TERF2IP Monoclonal specifically detects TERF2IP in Human samples. It is validated for Western Blot, ELISA.Specifications
TERF2IP | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
TERF2IP | |
Human TERF2IP | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, ELISA | |
RAP1 5G7/1 | |
Western Blot, ELISA | |
dopamine receptor interacting protein 5, Dopamine receptor-interacting protein 5, DRIP5, hRap1, RAP1 homolog, RAP1TERF2-interacting telomeric protein 1, Repressor/activator protein 1 homolog, telomeric repeat binding factor 2, interacting protein, telomeric repeat-binding factor 2-interacting protein 1, TRF2-interacting telomeric protein 1, TRF2-interacting telomeric RAP1 protein | |
Mouse | |
Protein A purified | |
Cellular Markers, Chromatin Research, DNA Repair, Endosome Markers, Lysosome Markers | |
54386 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction