Learn More
Tergitol(R) 15-S-7, Surfactant, Spectrum™ Chemical
SDP
Catalog No. 18607255 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Volume (Metric):
500 mL
Catalog No. 18-607-255
Supplier: Spectrum Chemical Manufacturing Corporation T1258500ML
T1258, 84133-50-6, C12-14H25-29O[CH2CH2O]xH
Spectrum™ Chemical Tergitol(R) 15-S-7, Surfactant is a Secondary Alcohol Ethoxylate surfactant and is an excellent emulsifier & detergent, enhances paint thickener efficiency and aids in rinse-ability of solvent-based systems. This product is suggested for use in cleaners, prewash spotters and paints & coatings.
Specifications
Volume (Metric) 500 mL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.