Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tergitol(R) NP-9, Surfactant, Spectrum™ Chemical

Click to view available options
Quantity:
100 mL
4 L
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical Tergitol(R) NP-9, Surfactant is a Nonylphenol Ethoxylate based surfactant. It is nonionic and provides excellent detergency, outstanding wetting, versatile solubility characteristics and low odor. This product is suggested for use in cleaners & detergents, prewash spotters, paints & coatings, dust control and metalworking fluids.
Specifications
Specifications
CAS Max % | 1 |
pH | 5.0 to 8.0 |
Quantity | 4 L |
Packaging | Amber Glass Bottle |
Specific Gravity | 1.042-1.062 |
Water | 0.003 |
Grade | Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction