Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tergitol(R) TMN-10, Surfactant, Spectrum™ Chemical
SDP

Catalog No. 18607284
Click to view available options
Volume (Metric):
100 mL

T1288, 60828-78-6, C12H25O(CH2CH2O)nH

Spectrum™ Chemical Tergitol(R) TMN-10, Surfactant is a Nonylphenol Ethoxylate. It is nonionic and provides excellent detergency, outstanding wetting, versatile solubility characteristics and low odor. This product is suggested for use in cleaners & degreasers, prewash spotters and oil field chemicals.

Specifications

pH 5 to 8
Composition C12H25O(CH2CH2O)nH
Volume (Metric) 100 mL
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.