Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tergitol(R) TMN-10, Surfactant, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Volume (Metric):
2.5 L
Description
Spectrum™ Chemical Tergitol(R) TMN-10, Surfactant is a Nonylphenol Ethoxylate. It is nonionic and provides excellent detergency, outstanding wetting, versatile solubility characteristics and low odor. This product is suggested for use in cleaners & degreasers, prewash spotters and oil field chemicals.
Specifications
Specifications
pH | 5 to 8 |
Composition | C12H25O(CH2CH2O)nH |
Volume (Metric) | 2.5 L |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction