Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tergitol(R) TMN-10, Surfactant, Spectrum™ Chemical
SDP

T1288, 60828-78-6, C12H25O(CH2CH2O)nH

Specifications

pH 5 to 8
Composition C12H25O(CH2CH2O)nH
View More Specs

Products 1
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
18-607-289
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
T12882.5LTPL
2.5 L N/A N/A N/A
Description

Description

Spectrum™ Chemical Tergitol(R) TMN-10, Surfactant is a Nonylphenol Ethoxylate. It is nonionic and provides excellent detergency, outstanding wetting, versatile solubility characteristics and low odor. This product is suggested for use in cleaners & degreasers, prewash spotters and oil field chemicals.

Specifications

Specifications

5 to 8
C12H25O(CH2CH2O)nH
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.