Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terminal uridylyltransferase 4 Polyclonal Antibody, Huabio

Catalog No. 502544665
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4665 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4665 Supplier HUABIO RESEARCH INC Supplier No. ER200133
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Terminal uridylyltransferase 4 Polyclonal antibody specifically detects Terminal uridylyltransferase 4 in Human samples. It is validated for Western Blot

Specifications

Antigen Terminal uridylyltransferase 4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene TUT4
Gene Accession No. Q5TAX3
Gene Alias 6030404K05Rik; 9230115F04Rik; Kiaa0191; mKIAA0191; PAP associated domain containing 3; PAPD3; PPAPD3; RP23-406B13.1; terminal uridylyl transferase 4; terminal uridylyltransferase 4; terminal uridylyltransferase 4-like; TUT4; TUTase 4; ZCCHC11; Zinc finger CCHC domain-containing protein 11; zinc finger CCHC-type containing 11; zinc finger, CCHC domain containing 11
Host Species Rabbit
Immunogen Recombinant protein within human terminal uridylyltransferase 4 aa 1-200.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23318
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.