Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(+)-Terpinen-4-ol, ≥98.5% (sum of enantiomers, GC), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
1 mL
5 mL
Description
Featured Industries: Food and Beverages

Specifications
Specifications
CAS | 2438-10-0 |
Density | 0.933 g/mL (at 20°C) |
Flash Point | 79°C - closed cup |
Physical Form | Neat |
Linear Formula | C10H18 O |
MDL Number | MFCD00077431 |
Boiling Point | 211°C to 213°C |
Synonym | (S)-1-Isopropyl-4-methyl-3-cyclohexen-1-ol; (S)-p-Menth-1-en-4-ol |
Type | Analytical Standard |
Quantity | 5 mL |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction