Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

γ-Terpinene, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187515
Click to view available options
Quantity:
1 mL
5 mL

γ-Terpinene is a monoterpene hydrocarbon present in essential oils extracted from Thymus vulgaris L., and exhibits antimicrobial, carminative and expectorant activities.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 99-85-4
Vapor Pressure ˜0.7 mmHg (20°C)
Vapor Density 4.7 (vs. air)
Density 0.85 g/mL (at 25°C)
Flash Point 50°C - closed cup
Physical Form Neat
Linear Formula C10H16
MDL Number MFCD00001537
Boiling Point 182°C
Synonym 1-Isopropyl-4-methyl-1,4-cyclohexadiene; p-Mentha-1,4-diene
Type Analytical Standard
Quantity 1 mL
Formula Weight 136.23
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.