Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Terpineol, Practical, Spectrum™ Chemical

Click to view available options
Quantity:
100 mL
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Max % | 1 |
Boiling Point | 213°C to 224°C |
Packaging | Amber Glass Bottle |
Refractive Index | 1.482-1.484 |
Quantity | 20 L |
Specific Gravity | 0.932 - 0.937 |
Grade | Practical |
Physical Form | Liquid |
Chemical Name or Material | Terpineol |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction