Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Terry Cloth Sweatband for Hard Hats

Catalog No. 19046446 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
One size
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-046-446 One size
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-046-446 Supplier PIP™ Supplier No. 39620870BGE
This item is not returnable. View return policy

PIP™ Terry Cloth Sweatband for Hard Hats helps absorb sweat at forehead

Attaches to hard hat suspension with plastic snaps

Specifications

Color Beige
For Use With (Equipment) Hard Hats
Material Terrycloth
Product Type Heat Stress
Size One Size
Width (Metric) 4.5 in.
Width (English) 11 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.