Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TERT Antibody (2C4), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 21 publication
$254.00 - $619.00
Specifications
Antigen | TERT |
---|---|
Clone | 2C4 |
Dilution | Western Blot 1:500, Chromatin Immunoprecipitation reported in scientific literature (PMID 30936423), Flow Cytometry 1:50 - 1:200, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200. Use reported in scientific literature (PMID 26725521), Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50, Chromatin Immunoprecipitation (ChIP) |
Applications | Western Blot, ChIP Assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100317SS
![]() |
Novus Biologicals
NB100317SS |
0.025 mL |
Each for $254.00
|
|
|||||
NB100317
![]() |
Novus Biologicals
NB100317 |
0.1 mL |
Each for $619.00
|
|
|||||
Description
TERT Monoclonal specifically detects TERT in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation (ChIP).Specifications
TERT | |
Western Blot 1:500, Chromatin Immunoprecipitation reported in scientific literature (PMID 30936423), Flow Cytometry 1:50 - 1:200, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200. Use reported in scientific literature (PMID 26725521), Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50, Chromatin Immunoprecipitation (ChIP) | |
Monoclonal | |
Ascites | |
RUO | |
Human, Mouse, Rat | |
O14746 | |
7015 | |
Full-length recombinant human Telomerase reverse transcriptase (TERT) from insect cells. [Uniprot: O14746]. | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
2C4 | |
Western Blot, ChIP Assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Autophagy, Cancer, Cellular Markers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Embryonic Stem Cell Markers, Mitophagy, Stem Cell Markers | |
PBS with 0.02% Sodium Azide | |
EST2Telomerase-associated protein 2, hEST2, TCS1EC 2.7.7.49, Telomerase catalytic subunit, telomerase reverse transcriptase, TP2HEST2, TRTEC 2.7.7 | |
TERT | |
IgM | |
Unpurified | |
127 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title