Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
tert-Butanol (Certified), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Poly Bottle
Specifications
Specifications
pH | 7 |
Melting Point | 25°C |
Acidity | 0.005% max. |
Density | 0.780g/cm³ |
Boiling Point | 83°C |
Quantity | 500 mL |
Identification | Pass Test |
Packaging | Poly Bottle |
Physical Form | Liquid |
Residue after Evaporation | 0.002% max. |
Show More |
Safety and Handling


Hazard Statement | DANGER! |
Precautionary Statement | Emergency Overview Flammable liquid and vapor. Irritating to eyes, respiratory system and skin. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Obtain medical attention. NFPA Health:2 Flammability:3 Instability:0 |
DOTInformation | DOT Class 3, : Flammable Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction