Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butoxydiphenylchlorosilane (stabilized with CaCO3) 95.0+%, TCI America™
SDP

Catalog No. B14365ML Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 mL
25 mL

Chemical Identifiers

CAS 17922-24-6
Molecular Formula C16H19ClOSi
Molecular Weight (g/mol) 290.862
MDL Number MFCD00077672
InChI Key MSZDOMFSWXWKTK-UHFFFAOYSA-N
Synonym tert-Butoxychlorodiphenylsilane, tert-Butoxydiphenylsilyl Chloride
PubChem CID 2735650
IUPAC Name chloro-[(2-methylpropan-2-yl)oxy]-diphenylsilane
SMILES CC(C)(C)O[Si](C1=CC=CC=C1)(C2=CC=CC=C2)Cl

Specifications

Color Colorless
Boiling Point 112°C
Quantity 5 mL
UN Number 2986
Formula Weight 290.86
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butoxydiphenylchlorosilane (stabilized with CaCO3)
TSCA No
Recommended Storage Freezer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.