Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl 5-Norbornene-2-carboxylate (endo- and exo- mixture) 95.0+%, TCI America™
SDP

Catalog No. B426725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B426725G 25 g
B42675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B426725G Supplier TCI America Supplier No. B426725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 154970-45-3
Molecular Formula C12H18O2
Molecular Weight (g/mol) 194.274
MDL Number MFCD18083704
InChI Key BZBMBZJUNPMEBD-UHFFFAOYSA-N
Synonym 5-Norbornene-2-carboxylic Acid tert-Butyl Ester, tert-Butyl Bicyclo[2.2.1]hept-5-ene-2-carboxylate, Bicyclo[2.2.1]hept-5-ene-2-carboxylic Acid tert-Butyl Ester
PubChem CID 15607334
IUPAC Name tert-butyl bicyclo[2.2.1]hept-2-ene-5-carboxylate
SMILES CC(C)(C)OC(=O)C1CC2CC1C=C2

Specifications

Color Yellow
Boiling Point 103°C
Quantity 25 g
Formula Weight 194.27
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyl 5-Norbornene-2-carboxylate (endo- and exo- mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.