Learn More
tert-Butyl alcohol, anhydrous, 99.5%, packaged under Argon in resealable ChemSeal™ bottles
Description
tert-Butyl alcohol is used as a solvent and an ethanol denaturant. It is an active ingredient in paint remover. It acts as an intermediate in the production of methyl tertiary butyl ether and ethyl tertiary butyl ether by reaction with methanol and ethanol. It is also used in the preparation of tert-butyl chloride, tert-butyl hydroperoxide, isobutylene and methyl methacrylate. It serves as a fuel additive as well as a gasoline octane booster.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon

Specifications
Specifications
Melting Point | 25°C |
Color | Colorless |
Density | 0.786 g/mL |
Boiling Point | 83°C |
Flash Point | 11°C (52°F) |
Assay Percent Range | 99.8% |
Packaging | ChemSeal™ bottles |
Refractive Index | 1.387 |
Linear Formula | (CH3)3COH |
Quantity | 250 mL |
Show More |
Safety and Handling
Hazard Category | H225-H319-H332-H335-H336 |
Hazard Statement | GHS H Statement H225-H332-H319-H335 Highly flammable liquid and vapor. Harmful if inhaled. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BUTANOLS |
EINECSNumber | 200-889-7 |
RTECSNumber | EO1925000 |
TSCA | Yes |
storageNote1 |
|
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.