Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl Alcohol, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. p-5391085
Click to view available options
Quantity:
500 mL
Packaging:
HDPE Pressure Bottom Poly Bottle

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 75-65-0
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
InChI Key DKGAVHZHDRPRBM-UHFFFAOYSA-N
Synonym tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
PubChem CID 6386
ChEBI CHEBI:45895
IUPAC Name 2-methylpropan-2-ol
SMILES CC(C)(C)O

Specifications

Flash Point 11.00°C
Packaging HDPE Pressure Bottom Poly Bottle
Quantity 500 mL
Specific Gravity 1L = 0.78kg
Formula Weight 74.12
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material tert-Butyl Alcohol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.