Learn More
tert-Butyl bromide, 98+%, stab. with potassium carbonate
Description
tert-Butyl bromide is used as a raw material in synthetic organic chemistry. It is used to study the massive deadenylation of adenine based-nucleosides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
tert-Butyl bromide is used as a raw material in synthetic organic chemistry. It is used to study the massive deadenylation of adenine based-nucleosides.
Solubility
Immiscible with water.
Notes
Incompatible with strong bases and strong oxidizing agents.
Specifications
Specifications
| Melting Point | °C to 20°C |
| Density | 1.210 |
| Boiling Point | 74°C to 76°C |
| Flash Point | 18°C (64°F) |
| Assay Percent Range | 98%, stabilized with potassium carbonate |
| Refractive Index | 1.428 |
| Quantity | 100 g |
| UN Number | UN2342 |
| Beilstein | 1730892 |
| Merck Index | 14,1555 |
| Show More |
Safety and Handling
| Hazard Category | H225-H315-H319-H335 |
| Hazard Statement | GHS H Statement H225 |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BROMOMETHYLPROPANES |
| EINECSNumber | 208-065-9 |
| RTECSNumber | TX4150000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.