Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl carbamate, 98%

Catalog No. AC173450050
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle

CAS: 4248-19-5 | C5H11NO2 | 117.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4248-19-5
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
InChI Key LFKDJXLFVYVEFG-UHFFFAOYSA-N
Synonym t-butyl carbamate, carbamic acid tert-butyl ester, boc amine, boc-amide, carbamic acid, 1,1-dimethylethyl ester, boc-nh2, tert-butylcarbamat, n-boc amine, boc-amine, tert butyl carbamate
PubChem CID 77922
IUPAC Name tert-butyl carbamate
SMILES CC(C)(C)OC(=O)N

Specifications

Melting Point 105.0°C to 109.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula H2NCO2C(CH3)3
Beilstein 03,II,26
Solubility Information Solubility in water: insoluble
Quantity 5 g
Formula Weight 117.15
Percent Purity 98%
Physical Form Needles
Chemical Name or Material tert-Butyl carbamate
Show More Show Less
missing translation for 'healthHazard1' Warning
EINECSNumber 224-209-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.