Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyloxonium tetrafluoroborate, 1M solution in methylene chloride

Catalog No. AC175110250
Click to view available options
Quantity:
25 mL
100 mL

CAS: 368-39-8 | C6H15BF4O | 189.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triethyloxonium tetrafluoroborate
Name Note 1M Solution in Methylene Chloride
CAS 368-39-8, 75-09-2
Molecular Formula C6H15BF4O
Linear Formula (C2H5)3OBF4
MDL Number MFCD00044423
Fieser 01,1210; 02,430; 03,303; 04,527; 05,691; 06,611; 07,386; 08,500; 09,482; 10,417; 15,337
Density 1.3280g/mL
Synonym triethyloxonium tetrafluoroborate, triethyloxonium fluoroborate, triethyloxonium fluoborate, triethyloxonium borofluoride, triethoxonium fluoroborate, meerwein's reagent, triethyloxidanium tetrafluoroborate, unii-z0b19dd36j, triethyloxonium tetraflouroborate, boron tetrafluoride triethyl oxonium
Solubility Information Solubility in water: reacts.
InChI Key IYDQMLLDOVRSJJ-UHFFFAOYSA-N
SMILES F[B-](F)(F)F.CC[O+](CC)CC
IUPAC Name tetrafluoroboranuide; triethyloxidanium
Molecular Weight (g/mol) 189.99
PubChem CID 2723982
Formula Weight 189.99
Packaging Plastic bottle
Specific Gravity 1.328
Quantity 25 mL
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.