Learn More
tert-Butyl carbazate, 98+%
Description
tert-Butyl carbazate is used in a palladium-catalyzed cross-coupling with vinyl halides to prepare N-Boc-N-alkenylhydrazines. It is utilized in solid phase peptide synthesis and in the optical purity determination of alfa-amino aldehyde. It reacts with aldehydes to get hydrazones, which finds application as an intermediate in the synthesis of HIV-1 protease inhibitors. In addition to this, it is used as a starting material for the preparation of BOC-azide, sulfonic and carboxylic hydrazides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
tert-Butyl carbazate is used in a palladium-catalyzed cross-coupling with vinyl halides to prepare N-Boc-N-alkenylhydrazines. It is utilized in solid phase peptide synthesis and in the optical purity determination of alfa-amino aldehyde. It reacts with aldehydes to get hydrazones, which finds application as an intermediate in the synthesis of HIV-1 protease inhibitors. In addition to this, it is used as a starting material for the preparation of BOC-azide, sulfonic and carboxylic hydrazides.
Solubility
Soluble in ethanol and methanol.
Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
Specifications
Specifications
| Melting Point | 37°C to 41°C |
| Boiling Point | 63°C to 65°C (0.1 mmHg) |
| Flash Point | 91°C (195°F) |
| Quantity | 100 g |
| UN Number | UN1325 |
| Beilstein | 1756967 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Soluble in ethanol and methanol. |
| Formula Weight | 132.16 |
| Percent Purity | ≥98% |
| Show More |
Safety and Handling
| Hazard Category | H228-H301+H311-H315-H319-H335 |
| Hazard Statement | GHS H Statement H228 Flammable solid. |
| Precautionary Statement | P210-P240-P241-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 4.1; Packaging Group: II |
| EINECSNumber | 212-795-3 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.