Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl diethylphosphonoacetate, 95%

Catalog No. p-7047116
Click to view available options
Quantity:
5 g
25 g

CAS: 27784-76-5 | C10H21O5P | 252.247 g/mol

tert-Butyl diethylphosphonoacetate is used as a reactant for preparation of hydroxymethylated dihydroxyvitamin D3 analogs via Wittig-Horner approach, as potential antitumor agents, synthesis of phosphopeptide mimetic prodrugs targeted to Src homology 2 (SH2) domain of signal transducer and activator of transcription 3 (Stat 3) and bicyclic triaminophosphine-promoted stereoselective synthesis of a,β-unsaturated esters, fluorides, and nitriles from aldehydes and ketones using Wadsworth-Emmons phosphonates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl diethylphosphonoacetate is used as a reactant for preparation of hydroxymethylated dihydroxyvitamin D3 analogs via Wittig-Horner approach, as potential antitumor agents, synthesis of phosphopeptide mimetic prodrugs targeted to Src homology 2 (SH2) domain of signal transducer and activator of transcription 3 (Stat 3) and bicyclic triaminophosphine-promoted stereoselective synthesis of a,ß-unsaturated esters, fluorides, and nitriles from aldehydes and ketones using Wadsworth-Emmons phosphonates

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27784-76-5
Molecular Formula C10H21O5P
Molecular Weight (g/mol) 252.247
MDL Number MFCD00075414
InChI Key NFEGNISFSSLEGU-UHFFFAOYSA-N
Synonym tert-butyl diethylphosphonoacetate, tert-butyl 2-diethoxyphosphoryl acetate, diethyl boc-methyl phosphonate, diethoxy-phosphoryl-acetic acid tert-butyl ester, t-butyl diethylphosphonoacetate, diethyl phosphonoacetic acid tert-butyl ester, t-butyl diethyl phosphonoacetate, diethylphosphonoacetic acid tert-butyl ester, diethyl 2-tert-butoxycarbonylmethyl phosphonate, acetic acid, diethoxyphosphinyl-, 1,1-dimethylethyl ester
PubChem CID 2773685
IUPAC Name tert-butyl 2-diethoxyphosphorylacetate
SMILES CCOP(=O)(CC(=O)OC(C)(C)C)OCC

Specifications

Density 1.074
Boiling Point 100°C to 103°C (1.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.431
Quantity 5 g
Beilstein 2050126
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 252.25
Percent Purity 95%
Chemical Name or Material tert-Butyl diethylphosphonoacetate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.