Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl Ethyl Ether 95.0+%, TCI America™
SDP

Catalog No. B154225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B154225ML 25 mL
B15425ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B154225ML Supplier TCI America Supplier No. B154225ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 637-92-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00009225
InChI Key NUMQCACRALPSHD-UHFFFAOYSA-N
Synonym tert-butyl ethyl ether, ethyl tert-butyl ether, etbe, propane, 2-ethoxy-2-methyl, ethyl tert-butyl oxide, ether, tert-butyl ethyl, 2-methyl-2-ethoxypropane, ethyl t-butyl ether, tert-butylethylether, methyl-2-ethoxypropane
PubChem CID 12512
IUPAC Name 2-ethoxy-2-methylpropane
SMILES CCOC(C)(C)C

Specifications

Color Colorless
Boiling Point 70°C
Quantity 25 mL
UN Number 1179
Formula Weight 102.18
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyl Ethyl Ether
EINECSNumber (2)-4058
RTECSNumber KN4730200
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.