Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich tert-Butyl isocyanide

Catalog No. 2606305G Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 g
5 g

Catalog No. 26-063-05G

Supplier: Sigma Aldrich 2606305G

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Carbon dative bond formation by tert-butyl isocyanide on the germanium (100) surface has been reported
  • It also forms complexes with metals and inserts into metal-carbon bonds to form iminoacyls.tert-butyl isocyanide was used in the synthesis of coumarines, 4h-chromenes and isoxazolines
  • It was also used to trap 2-cyclopropylidene-1,3-diones

Specifications

CAS 7188-38-7
Density 0.735 g/mL (at 25°C (literature))
Boiling Point 91°C (lit.)
Molecular Formula C5H9N
Refractive Index n20/D 1.376 (literature)
Linear Formula (CH3)3 CNC
MDL Number MFCD00000002
Quantity 5 g
Molecular Weight (g/mol) 83.13
Percent Purity 98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.