Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl methyl ether, HPLC Grade, 99+%

Catalog No. AA40477M1
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
4 x 4 L

Chromatography solvent | CAS: 1634-04-4 | C5H12O | 88.15 g/mol

Used as a Chromatography solvent. Acts as a fuel component in fuel for gasoline engines. They raise the oxygen content of gasoline(oxygenates).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1634-04-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00008812
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
Synonym tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
PubChem CID 15413
ChEBI CHEBI:27642
IUPAC Name 2-methoxy-2-methylpropane
SMILES CC(C)(C)OC

Specifications

Melting Point -109°C
Density 0.742 g/mL
Boiling Point 55°C to 56°C
Quantity 4 x 1 L
Flash Point -25°C (-13°F)
Odor Mild
Physical Form Liquid
Refractive Index 1.369
UN Number UN2350
Beilstein 1730942
Merck Index 14,6032
Solubility Information Slightly soluble in water. Highly soluble in ethanol and ethyl ether.
Formula Weight 88.15
Percent Purity ≥99%
Grade HPLC
Chemical Name or Material tert-Butyl methyl ether
Show More Show Less
Hazard Category H225-H315-H335
Hazard Statement GHS H Statement
H225-H315
Highly flammable liquid and vapor.
Causes skin irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BUTYL METHYL ETHER
EINECSNumber 216-653-1
RTECSNumber KN5250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.