Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl nitrite, 90%, pure

Catalog No. AC216221000
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy

Catalog No. AC216221000

Supplier: Thermo Scientific Chemicals 216221000

Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 540-80-7 | C4H9NO2 | 103.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-80-7
InChI Key IOGXOCVLYRDXLW-UHFFFAOYSA-N
Synonym t-butyl nitrite, 1,1-dimethylethyl nitrite, tert-butylnitrite, nitrous acid, 1,1-dimethylethyl ester, unii-4780h7u8lu, ccris 2098, alpha,alpha-dimethylethyl nitrite, nitrous acid, t-butyl ester, 2-methyl-2-propylnitrit, ch3 3cono
PubChem CID 10906
IUPAC Name tert-butyl nitrite
SMILES CC(C)(C)ON=O

Specifications

Color Yellow
Density 0.8600g/mL
Boiling Point 61.0°C to 63.0°C
Flash Point −10°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Molecular Formula C4H9NO2
Refractive Index 1.3670 to 1.3700
Linear Formula (CH3)3CNO2
Quantity 100 g
Beilstein 01,382
Fieser 07,48
Merck Index 15,1586
Specific Gravity 0.86
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,ether,chloroform and carbon,disulfide,practically insoluble in glycerol
Molecular Weight (g/mol) 103.12
Formula Weight 103.12
Percent Purity 90%
Grade Pure
Physical Form Liquid
Chemical Name or Material tert-Butyl nitrite
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 208-757-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.