Alfa Aesar™ tert-Butyl pivalate, 98%

Manufacturer: Alfa Aesar™ H5339522

Quantity 100g

 View more versions of this product

Catalog No. AAH5339522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 16474-43-4
    Molecular Formula C9H18O2
    Linear Formula (CH3)3CCO2C(CH3)3
    Formula Weight 158.24
    MDL Number MFCD01861974
    Synonym tert-Butyl trimethylacetate
    Chemical Name or Material tert-Butyl pivalate

    Specifications

    Formula Weight 158.24
    Molecular Formula C9H18O2
    Linear Formula (CH3)3CCO2C(CH3)3
    MDL Number MFCD01861974
    CAS 16474-43-4
    Synonym tert-Butyl trimethylacetate
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 134° to 135°C
    Flash Point >62°C (144°F)
    Quantity 100g
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Chemical Name or Material tert-Butyl pivalate
    Assay 98%

    Description

    Applications
    tert-Butyl pivalate is used as pharmaceutical intermediate.

    Notes
    Stable under recommended storage conditions. Store away from oxidizing agents.

    GHS H Statement
    H227
    Combustible liquid.

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit