Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl Propionate 98.0+%, TCI America™
SDP

Catalog No. B3327100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3327100G 100 g
B332725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B3327100G Supplier TCI America Supplier No. B3327100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20487-40-5
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00009304
InChI Key JAELLLITIZHOGQ-UHFFFAOYSA-N
Synonym tert-butyl propionate, propanoic acid, 1,1-dimethylethyl ester, propionic acid, tert-butyl ester, t-butyl propionate, t-butyl propanoate, tert-butylpropionate, 1,1-dimethylethyl propionate, tert-?butyl propionate, propionic acid tert-butyl ester, acmc-1cr5r
PubChem CID 88561
IUPAC Name tert-butyl propanoate
SMILES CCC(=O)OC(C)(C)C

Specifications

Color Colorless
Boiling Point 119°C
Quantity 100 g
UN Number 1914
Formula Weight 130.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyl Propionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.