Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl vinyl ether, 98%, stabilized, Acros Organics

Click to view available options
Quantity:
10mL
50mL
Packaging:
Glass bottle
926-02-3
C6H12O
100.16
MFCD00048246
PGYJSURPYAAOMM-UHFFFAOYSA-N
tert-butyl vinyl ether,2-ethenyloxy-2-methylpropane,propane, 2-ethenyloxy-2-methyl,propane,2-ethenyloxy-2-methyl,t-butyl vinyl ether,tert-butylvinyl ether,vinyltertiary-butylether,tertiarybutyl vinyl ether,vinyl tert.-butyl ether,tertiary butyl vinyl ether
70220
2-ethenoxy-2-methylpropane
CC(C)(C)OC=C
926-02-3
C6H12O
100.16
MFCD00048246
PGYJSURPYAAOMM-UHFFFAOYSA-N
tert-butyl vinyl ether,2-ethenyloxy-2-methylpropane,propane, 2-ethenyloxy-2-methyl,propane,2-ethenyloxy-2-methyl,t-butyl vinyl ether,tert-butylvinyl ether,vinyltertiary-butylether,tertiarybutyl vinyl ether,vinyl tert.-butyl ether,tertiary butyl vinyl ether
70220
2-ethenoxy-2-methylpropane
CC(C)(C)OC=C
Flash Point -17°C
Boiling Point 78°C
Specific Gravity 0.762
Viscosity 0.41 mPa.s (20°C)
Density 0.7620g/mL
Refractive Index 1.3970 to 1.399
Color Colorless to Yellow
Quantity 10mL
Infrared Spectrum Authentic
Packaging Glass bottle
Stabilizer approx. 100ppm KOH
Assay Percent Range 97.5% min. (GC)
Solubility Information Solubility in water: sparingly soluble
Formula Weight 100.16
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material tert-Butyl vinyl ether
Show More Show Less

GHS H Statement:
Harmful to aquatic life with long lasting effects.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger