Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butylacetic Acid, 98%

Catalog No. AC107770500
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle

CAS: 1070-83-3 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1070-83-3
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00002715
InChI Key MLMQPDHYNJCQAO-UHFFFAOYSA-N
Synonym 3,3-dimethylbutyric acid, tert-butylacetic acid, t-butylacetic acid, butanoic acid, 3,3-dimethyl, 3,3-dimethyl-butanoic acid, unii-q80k0tzq55, butyric acid, 3,3-dimethyl, 3,3-dimethyl-butyric acid, 3,3-dimethyl-n-butyric acid, tert-butyric acid
PubChem CID 14057
ChEBI CHEBI:38647
IUPAC Name 3,3-dimethylbutanoic acid
SMILES CC(C)(C)CC(=O)O

Specifications

Melting Point -8°C to 6°C
Density 0.9100g/mL
Boiling Point 185.0°C to 190.0°C
Flash Point 88°C
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4090 to 1.4110
Linear Formula (CH3)3CCH2CO2H
Quantity 50 mL
Beilstein 02, 337
Specific Gravity 0.91
Merck Index 15, 1540
Solubility Information Solubility in water: 19g/L (20 c)
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material tert-Butylacetic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-982-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.