Learn More
tert-Butylacetyl chloride, 98+%
Description
tert-Butylacetyl chloride is used an acyl chloride compound for proteomics research. It is also used as intermediate for important acylation reagents agrochemical and pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
tert-Butylacetyl chloride is used an acyl chloride compound for proteomics research. It is also used as intermediate for important acylation reagents agrochemical and pharmaceutical.
Solubility
Reacts with water.
Notes
Moisture Sensitive Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Density | 0.967 |
| Boiling Point | 128°C to 130°C |
| Flash Point | 21°C (69°F) |
| Odor | Acrid |
| Refractive Index | 1.423 |
| Quantity | 25 g |
| UN Number | UN2924 |
| Beilstein | 1740421 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Reacts with water. |
| Show More |
Safety and Handling
| Hazard Category | H225-H314-H335 |
| Hazard Statement | GHS H Statement H225-H314-H318 Highly flammable liquid and vapor. Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S. |
| EINECSNumber | 230-356-4 |
| TSCA | Yes |
| Recommended Storage | Keep cold; Store under Nitrogen |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.